missing translation for 'onlineSavingsMsg'
Learn More

Chloropentafluorobenzene, 98+%

Catalog No. AAA1538418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1538418 50 g
AAA1538409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1538418 Supplier Thermo Scientific Chemicals Supplier No. A1538418
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 344-07-0 | C6ClF5 | 202.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 344-07-0
Molecular Formula C6ClF5
Molecular Weight (g/mol) 202.51
MDL Number MFCD00000534
InChI Key KGCDGLXSBHJAHZ-UHFFFAOYSA-N
Synonym chloropentafluorobenzene, pentafluorochlorobenzene, benzene, chloropentafluoro, pentafluorophenyl chloride, chloroperfluorobenzene, ccris 2394, pentafluorochloro benzene, 1-chloro-2,3,4,5,6-pentafluoro-benzene, benzene, 1-chloro-2,3,4,5,6-pentafluoro, pentafluorophenylchloride
PubChem CID 9579
IUPAC Name 1-chloro-2,3,4,5,6-pentafluorobenzene
SMILES FC1=C(F)C(F)=C(Cl)C(F)=C1F

Specifications

Density 1.568
Boiling Point 116°C to 118°C
Refractive Index 1.424
Quantity 50 g
Beilstein 1819389
Formula Weight 202.51
Percent Purity ≥98%
Chemical Name or Material Chloropentafluorobenzene
Hazard Category H331
Precautionary Statement P261-P271-P304+P340-P311-P501c
EINECSNumber 206-450-6
RTECSNumber CZ1225500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only