missing translation for 'onlineSavingsMsg'
Learn More

Chloropentacarbonylrhenium(I), 98%

Catalog No. AA4209603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4209603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4209603 Supplier Thermo Scientific Chemicals Supplier No. 04209603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14099-01-5 | C5ClO5Re | 361.707 g/mol

Pentacarbonylchlororhenium(I) was used to study π-electron deficient complexes between tricarbonylhalorhenium and diimine ligands a well as to prepare [3]-ferrocenophane complexes. It has also been employed in preparation of a series of mixed-valence compounds of rhenium and iron. These compounds find application in the study of charge-transfer processes. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14099-01-5
Molecular Formula C5ClO5Re
Molecular Weight (g/mol) 361.707
MDL Number MFCD00013296
InChI Key JQUUAHKBIXPQAP-UHFFFAOYSA-M
Synonym pentacarbonylchlororhenium, carbon monoxide; chlororhenium, pentakis carbon monoxide ; chlororhenium, 5co.clre, rhenium i pentacarbonyl chloride, pentacarbonylchlororhenium i
PubChem CID 6096982
IUPAC Name carbon monoxide;chlororhenium
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].Cl[Re]

Specifications

Color Brown to White
Linear Formula Re(CO)5Cl
Quantity 1 g
UN Number UN3466
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 361.71
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Chloropentacarbonylrhenium(I)
Hazard Category H302+H312+H332-H315-H320-H335
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
EINECSNumber 237-948-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only