missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethyl)triethoxysilane 95.0+%, TCI America™

Numéro de catalogue. C140210G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C140210G 10 g
C140225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C140210G Fournisseur TCI America Code fournisseur C140210G
Il en reste null

Identifiants chimiques

CAS 15267-95-5
Molecular Formula C7H17ClO3Si
Molecular Weight (g/mol) 212.75
MDL Number MFCD00039354
InChI Key ZDOBWJOCPDIBRZ-UHFFFAOYSA-N
PubChem CID 84858
IUPAC Name (chloromethyl)triethoxysilane
SMILES CCO[Si](CCl)(OCC)OCC

Spécifications

Color Yellow
Boiling Point 180°C
Quantity 10 g
UN Number 1993
Formula Weight 212.75
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (Chloromethyl)triethoxysilane
EINECSNumber (2)-2079
RTECSNumber VV2275000
TSCA Yes