missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethyl)dimethylchlorosilane, 98%

Catalog No. AAA1489922
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1489922 100 g
AAA1489909 10 g
AAA1489914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1489922 Supplier Thermo Scientific Chemicals Supplier No. A1489922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1719-57-9 | C3H8Cl2Si | 143.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1719-57-9
Molecular Formula C3H8Cl2Si
Molecular Weight (g/mol) 143.08
MDL Number MFCD00000875
InChI Key ITKVLPYNJQOCPW-UHFFFAOYSA-N
Synonym chloro chloromethyl dimethylsilane, chloromethyl dimethylchlorosilane, chloromethyldimethylchlorosilane, silane, chloro chloromethyl dimethyl, cmdmcs, chloro cholromethyl dimethylsilane, ch3 2sicl ch2cl, chloromethylchlorodimethylsilane, dimethylchloromethylchlorosilane, chloro chloromethyl-dimethylsilane
PubChem CID 74393
IUPAC Name chloro-(chloromethyl)-dimethylsilane
SMILES C[Si](C)(Cl)CCl

Specifications

Boiling Point 110°C to 113°C
Quantity 100 g
Flash Point 21°C (69°F)
Odor Acrid
Refractive Index 1.437
Sensitivity Moisture sensitive
Density 1.082
Linear Formula ClCH2Si(CH3)2Cl
UN Number UN2985
Beilstein 1731933
Formula Weight 143.09
Percent Purity 98%
Chemical Name or Material (Chloromethyl)dimethylchlorosilane
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 217-006-6
RTECSNumber VV2095000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only