missing translation for 'onlineSavingsMsg'
Learn More

Chlorogenic acid

Catalog No. AAJ60457MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60457MD 250 mg
AAJ6045703 1 g
AAJ6045706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ60457MD Supplier Thermo Scientific Chemicals Supplier No. J60457MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An analog of caffeic acid | CAS: 327-97-9 | C16H18O9 | 354.31 g/mol

Chlorogenic acid is used as a food additive in coffee products, chewing gum and mint. It acts as an intermediate in the biosynthesis of lignin, which is an antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorogenic acid is used as a food additive in coffee products, chewing gum and mint. It acts as an intermediate in the biosynthesis of lignin, which is an antioxidant.

Solubility
Soluble in ethanol and acetone.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 327-97-9
Molecular Formula C16H18O9
Molecular Weight (g/mol) 354.31
MDL Number MFCD00003862
InChI Key CWVRJTMFETXNAD-JUHZACGLSA-N
Synonym chlorogenic acid, 3-caffeoylquinic acid, chlorogenate, 3-o-caffeoylquinic acid, 3-3,4-dihydroxycinnamoyl quinic acid, heriguard, hlorogenic acid, caffeoyl quinic acid, 3-caffeoylquinate, unii-318adp12ri
PubChem CID 1794427
ChEBI CHEBI:16112
IUPAC Name (1S,3R,4R,5R)-3-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy-1,4,5-trihydroxycyclohexane-1-carboxylic acid
SMILES O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O

Specifications

Melting Point 210°C (decomposition)
Quantity 250 mg
Merck Index 14,2142
Solubility Information Soluble in ethanol and acetone.
Formula Weight 354.31
Physical Form Powder
Chemical Name or Material Chlorogenic acid
EINECSNumber 206-325-6
RTECSNumber GU8480000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only