missing translation for 'onlineSavingsMsg'
Learn More

Chloroform (stabilized with Ethanol) 99.0+%, TCI America™

Catalog No. C0819500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0819500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C0819500ML Supplier TCI America Supplier No. C0819500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Physical Form Liquid
Color Colorless
Quantity 500 mL
Melting Point -64°C
Boiling Point 60°C
UN Number 1888
Formula Weight 119.37
Percent Purity ≥99.0% (GC)
Chemical Name or Material Chloroform (stabilized with Ethanol) [for HPLC Solvent]
EINECSNumber (2)-0037
RTECSNumber FS9100000
TSCA Yes