missing translation for 'onlineSavingsMsg'
Learn More

Chloroform/Methanol, 65:35, Ricca Chemical

Catalog No. R19991004C1
Change view
Click to view available options
Quantity:
4 L
Packaging:
Safety-coated Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R19991004C1 4 L Safety-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R19991004C1 Supplier Ricca Chemical Company Supplier No. R19991004C1
Only null left

Chemical Identifiers

CAS 67-66-3 , 67-56-1
Molecular Formula CHCl3
Molecular Weight (g/mol) Mixture
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

CAS Min % 30.0, 70.0
CAS Max % 40.0, 60.0
Quantity 4 L
Packaging Safety-coated Glass Bottle
Physical Form Liquid
Vapor Pressure 160mmHg@ 20°C (Chloroform), 97mmHg@20°C (Methanol)
Percent Purity 65/35
Grade Laboratory
Chemical Name or Material Chloroform/Methanol