missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethylpropylsilane 99.0+%, TCI America™

Catalog No. D15905ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15905ML 5 mL
D159025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15905ML Supplier TCI America Supplier No. D15905ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17477-29-1
Molecular Formula C5H13ClSi
Molecular Weight (g/mol) 136.694
MDL Number MFCD00039301
InChI Key HXVPUKPVLPTVCQ-UHFFFAOYSA-N
Synonym propyldimethylchlorosilane, chlorodimethylpropylsilane, n-propyldimethylchlorosilane, silane, chlorodimethylpropyl, dimethylpropylchlorosilane, chlorodimethyl propyl silane, chlorodimethyl-n-propylsilane
PubChem CID 87130
IUPAC Name chloro-dimethyl-propylsilane
SMILES CCC[Si](C)(C)Cl

Specifications

Color Colorless
Boiling Point 116°C
Quantity 5 mL
UN Number 2986
Formula Weight 136.69
Percent Purity ≥99.0% (GC)
Grade Reagent
Physical Form Liquid
Chemical Name or Material Chlorodimethylpropylsilane [Dimethylpropylsilylating Agent]
EINECSNumber (2)-2041
TSCA Yes