missing translation for 'onlineSavingsMsg'
Learn More

Chlorodiisopropylphosphine 98.0+%, TCI America™

Catalog No. C17205G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C17205G Supplier TCI America Supplier No. C17205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40244-90-4
Molecular Formula C6H14ClP
Molecular Weight (g/mol) 152.602
MDL Number MFCD00015027
InChI Key JZPDBTOWHLZQFC-UHFFFAOYSA-N
Synonym chlorodiisopropylphosphine, diisopropylchlorophosphine, di-i-propylchlorophosphine, phosphinous chloride, bis 1-methylethyl, ipr2pcl, pubchem6476, chlorodiisopropylphosphane, chloro diisopropylphosphine, acmc-209jc6, diisopropylphosphinyl chloride
PubChem CID 538967
IUPAC Name chloro-di(propan-2-yl)phosphane
SMILES CC(C)P(C(C)C)Cl

Specifications

Color Yellow
Boiling Point 158°C
Quantity 5 g
UN Number 2924
Formula Weight 152.60
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Chlorodiisopropylphosphine
TSCA No
Recommended Storage Refrigerator