missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene 98.0+%, TCI America™

Catalog No. C1948500G
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1948500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C1948500G Supplier TCI America Supplier No. C1948500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.556
MDL Number MFCD00000530
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
PubChem CID 7964
ChEBI CHEBI:28097
IUPAC Name chlorobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

Melting Point -45°C
Color Yellow
Quantity 500 g
Physical Form Clear Liquid at 20°C
UN Number 1134
Boiling Point 132°C
Formula Weight 112.56
Percent Purity ≥98.0% (GC)
Chemical Name or Material Chlorobenzene
EINECSNumber (3)-0031
RTECSNumber CZ0175000
TSCA Yes