missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, 99%

Catalog No. AAB21052AP
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB21052AP 500 mL
AAB210520F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB21052AP Supplier Thermo Scientific Chemicals Supplier No. B21052AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-90-7 | C6H5Cl | 112.556 g/mol

Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.

Solubility
Miscible with alcohol, chloroform, diethyl ether and benzene. Slightly immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.556
MDL Number MFCD00000530
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
PubChem CID 7964
ChEBI CHEBI:28097
IUPAC Name chlorobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

Melting Point -45°C
Density 1.106
Quantity 500 mL
Odor Aromatic
Refractive Index 1.524
UN Number UN1134
Beilstein 605632
Merck Index 14,2121
Boiling Point 132°C
Solubility Information Miscible with alcohol,chloroform,diethyl ether and benzene. Slightly immiscible with water.
Formula Weight 112.56
Percent Purity 99%
Chemical Name or Material Chlorobenzene
Show More Show Less
Hazard Category H226-H315-H332
Hazard Statement GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZENE
EINECSNumber 203-628-5
RTECSNumber CZ0175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only