missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, 99.5% min., ACS, MilliporeSigma™

Catalog No. MCX085530
Change view
Click to view available options
Quantity:
500 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MCX085530 500 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MCX085530 Supplier Technidata Supplier No. CX085530

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.556
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
PubChem CID 7964
ChEBI CHEBI:28097
IUPAC Name chlorobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

Color Colorless
Density 1.11g/cm3 (20°C)
Quantity 500 lb.
Explosion Limit 1.3 to 11%(V)
Physical Form Liquid
Assay Percent Range 99.5% min.
Linear Formula C6H5Cl
Vapor Pressure 12 hPa (20°C)
Formula Weight 112.56
Chemical Name or Material Chlorobenzene, For Synthesis
Recommended Storage Store at +15°C to +25°C