missing translation for 'onlineSavingsMsg'
Learn More

chloroacetic anhydride, 97%, Thermo Scientific™

Catalog No. AC404422500
Change view
Click to view available options
250g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC404422500 250g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC404422500 Supplier Thermo Scientific Chemicals Supplier No. 404422500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 541-88-8
Boiling Point 203°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C4H4Cl2O3
Linear Formula (ClCH2CO)2O
Quantity 250g
MDL Number MFCD00000929
Beilstein 02, 199
Fieser 01,129
Merck Index 15, 2113
Synonym chloroacetic anhydride, chloracetic anhydride, monochloroacetic acid anhydride, 2-chloroacetic anhydride, chloroacetic acid anhydride, chloroacetyl anhydride, acetic acid, chloro-, anhydride, unii-8q7txh7r5f, monochloroacetic anhydride, 8q7txh7r5f
Solubility Information Solubility in water: decomposes. Other solubilities: freely soluble in ether and chloroform,slightly soluble in benzene,practically insoluble in cold ligroin
InChI Key PNVPNXKRAUBJGW-UHFFFAOYSA-N
SMILES ClCC(=O)OC(=O)CCl
IUPAC Name (2-chloroacetyl) 2-chloroacetate
Molecular Weight (g/mol) 170.97
PubChem CID 10946
Formula Weight 170.98
Percent Purity 97%
Chemical Name or Material chloroacetic anhydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 208-794-2

RUO – Research Use Only