missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetic Acid, Crystal, ACS, 99%, Spectrum™ Chemical

Catalog No. 18609686
Change view
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609686 500 g Amber Glass Bottle
18609687 2.5 kg Amber Glass Bottle
18609688 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18609686 Supplier Spectrum Chemical Mfg Cor Supplier No. M1320500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 79-11-8
Molecular Formula C2H3ClO2
Molecular Weight (g/mol) 94.49
InChI Key FOCAUTSVDIKZOP-UHFFFAOYSA-N
IUPAC Name 2-chloroacetic acid
SMILES OC(=O)CCl

Specifications

CAS Max % 1
Ignition Residue 0.0002
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 0.99
Grade ACS
Insoluble Matter 0.0001

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.