missing translation for 'onlineSavingsMsg'
Learn More

Chloro(triphenylphosphine)gold(I), Premion™, 99.99% (metals basis), Au 39.3% min

Catalog No. AA4100706
Change view
Click to view available options
Quantity:
0.5 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4100706 5 g
AA4100702 0.5 g
AA4100703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4100706 Supplier Thermo Scientific Chemicals Supplier No. 04100706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14243-64-2 | C18H15AuClP | 494.71 g/mol

Chloro(triphenylphosphine)gold(I) is a common reagent in gold chemistry. It catalyzes certain rearrangement reactions in organic synthesis. It is also used to prepare 1-methylthyminato-N3-triphenylphosphinegold(I) by reacting with 1-methylthymine. It acts as a precursor for the synthesis of gold(I) and gold(III) organometallic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14243-64-2
Molecular Formula C18H15AuClP
Molecular Weight (g/mol) 494.71
MDL Number MFCD00009588
InChI Key IFPWCRBNZXUWGC-UHFFFAOYSA-M
Synonym chloro triphenylphosphine gold i, chloro triphenylphosphine gold, chlorotriphenylphosphine gold i, triphenylphosphinegold i chloride, aucl pph3, pubchem16017, ph3p aucl, chlorogold; triphenylphosphane, chlorogold; triphenylphosphine, chloranylgold; triphenylphosphane
PubChem CID 10874691
IUPAC Name chlorogold;triphenylphosphane
SMILES Cl[Au].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Odor Odorless
Assay Percent Range 99.99% (Metal basis, Au 39.3% min.)
Quantity 5 g
Solubility Information Soluble in methylene chloride,acetonitrile,benzene and acetone. Insoluble in water and ethanol.
Formula Weight 494.71
Physical Form Powder
Chemical Name or Material Chloro(triphenylphosphine)gold(I)
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-117-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only