missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloro-N,N,N',N'-tetramethylformamidinium Hexafluorophosphate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 107°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 280.58 |
| Percent Purity | ≥98.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Chloro-N,N,N′,N′-tetramethylformamidinium Hexafluorophosphate |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |