missing translation for 'onlineSavingsMsg'
Learn More

Chlorine in Heavy Mineral Oil standard solution, Specpure™ 100^mg/g (0.010%)

 

Supplier:  Thermo Scientific Chemicals 040549AE

Catalog No. AA40549AE


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Chlorine in Heavy Mineral Oil standard solution
MFCD02264398
Specpure™
100μg/g (0.010%)
100 mL

Chemical Identifiers

MFCD02264398
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350
May cause cancer.

P301+P310-P331-P501c

H304

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only