missing translation for 'onlineSavingsMsg'
Learn More

Chloride Standard, (NaCl in Water), Certified, 500ppm (1mL = 0.5mg Cl-) (0.0141N), LabChem™

Catalog No. LC130101
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC130101 500 mL Poly Bottle
LC130102 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC130101 Supplier LabChem Supplier No. LC130101

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Packaging Poly Bottle
CAS Max % 99.92, 0.08
Identification Passes Test
Physical Form Liquid
Concentration 500 ppm (1mL = 0.5mg Cl-) (0.0141N)
Linear Formula NaCl
Color Colorless
Solubility Soluble in water
Quantity 500 mL
Formula Weight 58.44
Grade Certified
Chemical Name or Material Chloride Standard, 500ppm (1mL = 0.5mg Cl-) (0.0141N) (NaCl in Water)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature