missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloride Standard, (NaCl in Water), Certified, 1000ppm ±5ppm (1mL = 1mg Cl-) (0.0282N), LabChem™
Specifications
Specifications
| Packaging | Poly Bottle |
| CAS Max % | 99.84, 0.16 |
| Identification | Passes Test |
| Physical Form | Liquid |
| Concentration | 1000 ppm ±5 ppm (1mL = 1mg Cl-) (0.0282N) |
| Linear Formula | NaCl |
| Color | Colorless |
| Solubility | Soluble in water |
| Quantity | 125 mL |
| Formula Weight | 58.44 |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Solution is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |