missing translation for 'onlineSavingsMsg'
Learn More

Chloride Standard, (NaCl in Water), Certified, 1000ppm ±5ppm (1mL = 1mg Cl-) (0.0282N), LabChem™

Catalog No. LC130007
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC130007 125 mL Poly Bottle
LC130001 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC130007 Supplier LabChem Supplier No. LC130007

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Packaging Poly Bottle
CAS Max % 99.84, 0.16
Identification Passes Test
Physical Form Liquid
Concentration 1000 ppm ±5 ppm (1mL = 1mg Cl-) (0.0282N)
Linear Formula NaCl
Color Colorless
Solubility Soluble in water
Quantity 125 mL
Formula Weight 58.44
Grade Certified
Chemical Name or Material Chloride Standard, 1000ppm (1mL = 1mg Cl-) (0.0282N) (NaCl in Water)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature