missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloride Standard, 500 ppm Cl- (0.0141 N), Ricca Chemical
Supplier: Ricca Chemical Company 195016
Specifications
| Natural Poly Bottle | |
| 98.0,0.08 | |
| 1 mL = 0.5 mg Cl | |
| ClNa | |
| Colorless | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 500 mL | |
| CHEBI:26710 |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| 5234 | |
| sodium chloride |