missing translation for 'onlineSavingsMsg'
Learn More

Chloride Standard, 100 ppm Cl-, Ricca Chemical

Catalog No. 19474
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
19474 120 mL Natural Poly Bottle
194716 500 mL Natural Poly Bottle
194732 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19474 Supplier Ricca Chemical Company Supplier No. 19474

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.01
CAS Max % 100.0, 0.02
Equivalence 1 mL = 0.1 mg Cl
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 120 mL
Chemical Name or Material Chloride Standard