missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloride Standard, 1000 ppm Cl- (0.0282 N), Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| Concentration or Composition Notes | 1000ppm |
| CAS Min % | 98.0, 0.16 |
| CAS Max % | 100.0, 0.17 |
| Equivalence | 1 mL = 1 mg Cl |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| Quantity | 120 mL |
| Grade | Not Available |
| Show More |