missing translation for 'onlineSavingsMsg'
Learn More

Chloride ISA, 0.100 M Sodium Chloride, Fisherbrand

Catalog No. ACCU0818500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACCU0818500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ACCU0818500 Supplier Ricca Chemical Company Supplier No. ACCU0818500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
Solubility Miscible with water
Solution Type Calibration Solution
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Chloride ISA, 0.1M Sodium Chloride
Show More Show Less