missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine digluconate Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019563
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019563 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019563 Supplier Merck Emd Millipore Supplier No. PHR12941G

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 18472-51-0
MDL Number MFCD00083599
Synonym Chlorhexidine digluconate solution; 1,6-Bis(N5-[p-chlorophenyl]-N1-biguanido)hexane; 1,1 '-Hexamethylenebis(5-[p-chlorophenyl]biguanide)
Quantity 1 g
Grade Certified Reference Material