missing translation for 'onlineSavingsMsg'
Learn More

Chlorendic anhydride, 95%, may cont. up to 3% chlorendic acid

Catalog No. AAL0734930
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0734930 250 g
AAL0734918 50 g
AAL073490B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0734930 Supplier Thermo Scientific Chemicals Supplier No. L0734930
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 115-27-5 | C9H2Cl6O3 | 370.812 g/mol

Chlorendic anhydride is used in the synthesis of polymers, used in production of fiberglass-reinforced resins for chemical industry equipment. It can be used to make alkyd resins for use in special inks and paints. It is used as a hardening agent in epoxy resins used in manufacture of printed circuit boards

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorendic anhydride is used in the synthesis of polymers, used in production of fiberglass-reinforced resins for chemical industry equipment. It can be used to make alkyd resins for use in special inks and paints. It is used as a hardening agent in epoxy resins used in manufacture of printed circuit boards

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 115-27-5
Molecular Formula C9H2Cl6O3
Molecular Weight (g/mol) 370.812
MDL Number MFCD00080438
InChI Key FLBJFXNAEMSXGL-UHFFFAOYSA-N
Synonym chlorendic anhydride, het anhydride, chloran 542, 4,7-methanoisobenzofuran-1,3-dione, 4,5,6,7,8,8-hexachloro-3a,4,7,7a-tetrahydro, 1,4,5,6,7,7-hexachloro-5-norbornene-2,3-dicarboxylic anhydride, chlorendic anhydride van, 4,5,6,7,8,8-hexachloro-3a,4,7,7a-tetrahydro-4,7-methanoisobenzofuran-1,3-dione, hexachloroendomethylene tetrahydrophthalic anhydride, hexachloro-5-norbornene-2,3-dicarboxylic anhydride, 5-norbornene-2,3-dicarboxylic anhydride, 1,4,5,6,7,7-hexachloro
PubChem CID 8265
SMILES C12C(C(=O)OC1=O)C3(C(=C(C2(C3(Cl)Cl)Cl)Cl)Cl)Cl

Specifications

Melting Point 235°C to 239°C
Density 1.73
Quantity 250 g
Beilstein 92693
Merck Index 14,2086
Solubility Information Insoluble in water.
Formula Weight 370.83
Percent Purity 96%
Assay may cont. up to 3% chlorendic acid
Chemical Name or Material Chlorendic anhydride
Hazard Category H315-H319-H335-H350
Hazard Statement GHS H Statement
H350-H315-H319-H335
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 204-077-3
RTECSNumber RB9080000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only