missing translation for 'onlineSavingsMsg'
Learn More

Chlorendic Acid 98.0+%, TCI America™

Catalog No. H005425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H005425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H005425G Supplier TCI America Supplier No. H005425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 115-28-6
Molecular Formula C9H4Cl6O4
Molecular Weight (g/mol) 388.827
MDL Number MFCD00167589
InChI Key DJKGDNKYTKCJKD-UHFFFAOYSA-N
Synonym Het Acid, 1,4,5,6,7,7-Hexachlorobicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic Acid, Hexachloro-endo-methylenetetrahydrophthalic Acid
PubChem CID 8266
ChEBI CHEBI:76603
IUPAC Name 1,2,3,4,7,7-hexachlorobicyclo[2.2.1]hept-2-ene-5,6-dicarboxylic acid
SMILES C1(C(C2(C(=C(C1(C2(Cl)Cl)Cl)Cl)Cl)Cl)C(=O)O)C(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 388.83
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chlorendic Acid
EINECSNumber (4)-0619
RTECSNumber RB9000000
TSCA Yes