missing translation for 'onlineSavingsMsg'
Learn More

Chloranilic Acid 98.0+%, TCI America™

Catalog No. C007725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C007725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C007725G Supplier TCI America Supplier No. C007725G
Only null left

Chemical Identifiers

CAS 87-88-7
Molecular Formula C6H2Cl2O4
Molecular Weight (g/mol) 208.978
MDL Number MFCD00001596
InChI Key IPPWILKGXFOXHO-UHFFFAOYSA-N
Synonym chloranilic acid, 2,5-dichloro-3,6-dihydroxy-p-benzoquinone, p-chloranilic acid, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro-3,6-dihydroxy, unii-yj8l3bb7y4, 2,5-dichloro-3,6-dihydroxybenzoquinone, 2,5-dihydroxy-3,6-dichlorobenzoquinone, yj8l3bb7y4, p-quinone, 2,5-dichloro-3,6-dihydroxy, p-benzoquinone, 2,5-dichloro-3,6-dihydroxy
PubChem CID 66604
IUPAC Name 2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione
SMILES C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O

Specifications

Melting Point 284°C
Color Red-Yellow
Quantity 25 g
Formula Weight 208.98
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Chloranilic Acid
RTECSNumber DK4005000
TSCA Yes