missing translation for 'onlineSavingsMsg'
Learn More

Chloramphenicol Crystalline MP Biomedicals

Catalog No. ICN19032105
Change view
Click to view available options
Quantity:
5 g
25 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19032105 5 g
ICN19032125 25 g
ICN19032190 500 g
ICN19032191 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19032105 Supplier MP Biomedicals Inc Supplier No. 0219032105

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56-75-7
Molecular Formula C11H12Cl2N2O5
Molecular Weight (g/mol) 323.126
InChI Key WIIZWVCIJKGZOK-RKDXNWHRSA-N
Synonym chloramphenicol, chlornitromycin, chloromycetin, levomycetin, chlorocid, halomycetin, levomicetina, alficetyn, fenicol, globenicol
PubChem CID 5959
ChEBI CHEBI:17698
IUPAC Name 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
SMILES C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-]

Specifications

Density 1.586 g/mL
Quantity 5 g
Formula Weight 323.15
Physical Form Crystalline
Chemical Name or Material Chloramphenicol