missing translation for 'onlineSavingsMsg'
Learn More

Chloramphenicol, 99.78%, MP Biomedicals™

Catalog No. ICN19032180
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19032180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19032180 Supplier MP Biomedicals Inc Supplier No. 0219032180

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56-75-7
Molecular Formula C11H12Cl2N2O5
Molecular Weight (g/mol) 323.126
InChI Key WIIZWVCIJKGZOK-RKDXNWHRSA-N
Synonym chloramphenicol, chlornitromycin, chloromycetin, levomycetin, chlorocid, halomycetin, levomicetina, alficetyn, fenicol, globenicol
PubChem CID 5959
ChEBI CHEBI:17698
IUPAC Name 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
SMILES C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-]

Specifications

Color White to Cream
Density 1.586g/mL
Assay Percent Range 0.9978
Quantity 100 g
Formula Weight 323.15
Physical Form Crystalline Powder
Chemical Name or Material Chloramphenicol