missing translation for 'onlineSavingsMsg'
Learn More

Chloramine-T trihydrate, 98%

Catalog No. AAA1204430
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1204430 250 g
AAA120440B 1000 g
AAA120440I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1204430 Supplier Thermo Scientific Chemicals Supplier No. A1204430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7080-50-4 | C7H13ClNNaO5S | 281.68 g/mol

Chloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).

Solubility
Soluble in water and ethyl alcohol. Insoluble in benzene and ethers.

Notes
Incompatible with strong oxidizing agents, acids and ammonia.

Chemical Identifiers

CAS 7080-50-4
Molecular Formula C7H13ClNNaO5S
Molecular Weight (g/mol) 281.68
MDL Number MFCD00149066
InChI Key RNROOTWAJBXJBD-UHFFFAOYSA-M
Synonym benzenesulfonamide, n-chloro-4-methyl-, sodium salt, trihydrate
PubChem CID 122197679
IUPAC Name N-chloro-4-methylbenzenesulfonamide;sodium;trihydrate
SMILES O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl

Specifications

Melting Point 167°C to 170°C
Quantity 250 g
Assay Percent Range 98%
UN Number UN3263
Beilstein 3924168
Flash Point 192°C (377°F)
Merck Index 14,2075
Solubility Information Soluble in water and ethyl alcohol. Insoluble in benzene and ethers.
Formula Weight 281.69 (227.67 Anhydrous)
Percent Purity 98%
Sensitivity Moisture sensitive
Chemical Name or Material Chloramine-T Trihydrate
Show More Show Less
Hazard Category H302-H314-H334-H335
Hazard Statement GHS H Statement
H334-H314-H318-H302
Precautionary Statement P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
EINECSNumber 204-854-7
RTECSNumber XT5616800
TSCA Yes
storageNote1 Moisture Sensitive
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only