Learn More
Chloramine-T trihydrate, 97+%
Chloramine-T trihydrate, >-97%, C7H14ClNNaO5S, CAS Number-7080-50-4, 5kg, 97+%, White, 192 deg.C, 281.69, GHS Signal Word: Danger | CAS: 7080-50-4 | C7H13ClNNaO5S | 281.68 g/mol
$66.75 - $1,524.15
Chemical Identifiers
| CAS | 7080-50-4 |
|---|---|
| Molecular Formula | C7H13ClNNaO5S |
| Molecular Weight (g/mol) | 281.68 |
| MDL Number | MFCD00149066 |
| InChI Key | RNROOTWAJBXJBD-UHFFFAOYSA-M |
| PubChem CID | 122197679 |
| SMILES | O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC227850250
|
N/A | 25 g | Plastic Bottle |
Each for $66.75
|
|
||||
|
AC227852500
|
N/A | 250 g | Plastic Bottle |
Each for $157.52
|
|
||||
|
AC227850010
|
N/A | 1 kg | Plastic Bottle |
Each for $347.74
|
|
||||
|
AC227850050
|
N/A | 5 kg | Plastic Drum |
Each for $1,524.15
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagentChemical Identifiers
| 7080-50-4 | |
| 281.68 | |
| RNROOTWAJBXJBD-UHFFFAOYSA-M | |
| O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl |
| C7H13ClNNaO5S | |
| MFCD00149066 | |
| 122197679 |
Specifications
| Chloramine T Trihydrate | |
| 7080-50-4 | |
| 170.0°C to 177.0°C | |
| White | |
| 97+% | |
| CH3C6H4SO2N(Cl)Na·3H2O | |
| 15, 2075 | |
| O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl | |
| 281.68 | |
| 281.69 | |
| Chloramine-T trihydrate |
| Crystalline Powder | |
| 8 to 10.3 (5% soln.) | |
| Authentic | |
| 192°C | |
| C7H13ClNNaO5S | |
| MFCD00149066 | |
| RNROOTWAJBXJBD-UHFFFAOYSA-M | |
| N-chloro-4-methylbenzenesulfonamide;sodium;trihydrate | |
| 122197679 | |
| ≥97% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Contact with acids liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only