missing translation for 'onlineSavingsMsg'
Learn More

Chloramine-T trihydrate, 97+%

Catalog No. AC227850250
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227850250 25 g Plastic Bottle
AC227852500 250 g Plastic Bottle
AC227850010 1 kg Plastic Bottle
AC227850050 5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC227850250 Supplier Thermo Scientific Chemicals Supplier No. 227850250

Chloramine-T trihydrate, >-97%, C7H14ClNNaO5S, CAS Number-7080-50-4, 5kg, 97+%, White, 192 deg.C, 281.69, GHS Signal Word: Danger | CAS: 7080-50-4 | C7H13ClNNaO5S | 281.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent

Chemical Identifiers

CAS 7080-50-4
Molecular Formula C7H13ClNNaO5S
Molecular Weight (g/mol) 281.68
MDL Number MFCD00149066
InChI Key RNROOTWAJBXJBD-UHFFFAOYSA-M
PubChem CID 122197679
SMILES O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl

Specifications

Type Chloramine T Trihydrate
Physical Form Crystalline Powder
pH 8 to 10.3 (5% soln.)
Melting Point 170.0°C to 177.0°C
Infrared Spectrum Authentic
Packaging Plastic Bottle
Color White
Quantity 25 g
Flash Point 192°C
Assay Percent Range 97+%
Linear Formula CH3C6H4SO2N(Cl)Na·3H2O
Merck Index 15, 2075
IUPAC Name N-chloro-4-methylbenzenesulfonamide;sodium;trihydrate
Formula Weight 281.69
Percent Purity ≥97%
Chemical Name or Material Chloramine-T trihydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Contact with acids liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only