missing translation for 'onlineSavingsMsg'
Learn More

Chloramine-T trihydrate, 97+%

Chloramine-T trihydrate, >-97%, C7H14ClNNaO5S, CAS Number-7080-50-4, 5kg, 97+%, White, 192 deg.C, 281.69, GHS Signal Word: Danger | CAS: 7080-50-4 | C7H13ClNNaO5S | 281.68 g/mol

Supplier:  Thermo Scientific Chemicals 227850050

Quantity 5 kg
Packaging Plastic Drum

 View more versions of this product

Catalog No. AC227850050


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
Specifications

Chemical Identifiers

7080-50-4
281.68
RNROOTWAJBXJBD-UHFFFAOYSA-M
O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl
C7H13ClNNaO5S
MFCD00149066
122197679

Specifications

Chloramine T Trihydrate
Crystalline Powder
8 to 10.3 (5% soln.)
Authentic
White
192°C
C7H13ClNNaO5S
MFCD00149066
RNROOTWAJBXJBD-UHFFFAOYSA-M
sodium (chloroimino)(4-methylphenyl)-λ⁶-sulfanoylolate trihydrate
122197679
≥97%
Chloramine-T trihydrate
7080-50-4
170.0°C to 177.0°C
Plastic Drum
5 kg
97+%
CH3C6H4SO2N(Cl)Na·3H2O
15, 2075
O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl
281.68
281.69
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Contact with acids liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Promotions

Promotions