missing translation for 'onlineSavingsMsg'
Learn More

Chitin, Practical grade, MP Biomedicals™

Catalog No. ICN10133491
Change view
Click to view available options
Quantity:
10 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10133491 1 kg
ICN10133410 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10133491 Supplier MP Biomedicals Inc Supplier No. 0210133491

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1398-61-4
Molecular Formula (C8H13NO5)nC16H28N2O11
Synonym chitin from shrimp shells, n-2r-2,4,5-trihydroxy-6-hydroxymethyl oxan-3-yl acetamide, chitin from shrimp shells, practical grade, powder, chitin from shrimp shells, vetec tm reagent grade, chitin from shrimp shells, practical grade, coarse flakes, chitin from shrimp shells, bioreagent, suitable for analysis of chitinase, purified powder

Specifications

Color White to Brown
Quantity 1 kg
Formula Weight 770.8
Grade Practical
Physical Form Powder
Chemical Name or Material Chitin