missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ CHIRALPAK™ CBH HPLC Guard Column, 5 μm Particle Size

Catalog No. 111008418
Change view
Click to view available options
Diameter (Metric) Inner:
3 mm
4 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
111008418 3 mm
111007950 4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111008418 Supplier MilliporeSigma™ Supelco™ Supplier No. 58558AST

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHIRALPAK™ CBH phase has cellobiohydrolase as the chiral selector immobilized on spherical 5 μm silica particles.

CHIRALPAK™ CBH phase has cellobiohydrolase as the chiral selector immobilized on spherical 5 μm silica particles. Used in reversed-phase mode, retention and enantioselectivity is regulated by changes of pH, buffer concentration and organic modifier. The mobile phases are mixtures of phosphate or acetate buffers and organic solvents such as 2-propanol or acetonitrile. The column is preferably used for the separation of enantiomers of basic drugs, particularly compounds containing one or more nitrogen atoms along with one or more hydrogen-bonding groups (alcohol, phenol, carbonyl, amide, ether, ester, etc.). Features: Direct reversed phase resolution of chiral molecules. Stable with a variety of organic modifiers. Available in analytical and semi-preparative sizes.

Specifications

Quantity 2/Pk.
Particle Size 5 μm
Length (Metric) 1 cm
Type Chiral HPLC Guard Column
Diameter (Metric) Inner 3 mm
Phase Chiral
For Use With (Application) HPLC
Product Line CHIRALPAK

CHIRALPAK is a registered trademark of Daicel Corp.