missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chicken anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody
Description
Chicken anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody for ELISA
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spécifications
Spécifications
| Antigène | Rat IgG (H+L) Cross-Adsorbed |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | >4.5 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 0.05% sodium azide; pH 7.2 |
| Espèces hôtes | Chicken |
| Immunogène | Gamma Immunoglobins Heavy and Light chains. |
| Méthode de purification | Affinity chromatography |
| Quantité | 1 mg |
| Afficher plus de résultats |