missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken Mouse IgG Chicken anti-Mouse, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Chicken anti-Mouse IgG Antibody, HRP conjugate detects level of Chicken Mouse IgG and has been published and validated for use in ELISA and western blotting.
Spécifications
Spécifications
| Antigène | Chicken anti-Mouse IgG |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugué | HRP |
| Formule | Affinity purified immunoglobulin conjugated with horseradish peroxidase. Liquid in 50mM HEPES, pH 7.0, 0.1M NaCl, 0.2% BSA and 0.1% Kathon CG. |
| Espèces hôtes | Chicken |
| Immunogène | Mouse IgG |
| Méthode de purification | Affinity Purified |
| Quantité | 1 mL |
| Primaire ou secondaire | Secondary |
| Afficher plus de résultats |
For Research Use Only