missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chicken anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody

Numéro de catalogue. A15971
Encompass
Modifier l'affichage
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
A15971 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. A15971 Fournisseur Invitrogen™ Code fournisseur A15971

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Chicken Polyclonal Secondary Antibody

Chicken anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody for ELISA

Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Spécifications

Antigène Goat IgG (H+L) Cross-Adsorbed
Applications ELISA
Classification Polyclonal
Concentration >4.5 mg/mL
Conjugué Unconjugated
Formule PBS with 0.05% sodium azide; pH 7.2
Espèces hôtes Chicken
Immunogène Gamma Immunoglobins Heavy and Light chains.
Méthode de purification Affinity chromatography
Quantité 1 mg
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Goat
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Secondary Antibody
Forme Liquid
Isotype IgY
Afficher plus de résultats Afficher moins de résultats