missing translation for 'onlineSavingsMsg'
Learn More

Chenodeoxycholic Acid, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022966
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022966 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022966 Supplier Merck Emd Millipore Supplier No. BP926

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 474-25-9
Melting Point 165°C to 167°C (literature)
Molecular Formula C24H40O4
Linear Formula C24H40O4
MDL Number MFCD00064142
Quantity 25 mg
Synonym 3 alpha,7 alpha-Dihydroxy-5 beta-cholanic acid; 5 beta-Cholanic acid-3 alpha,7 alpha-diol; Chenodiol
Formula Weight 392.57
Grade British Pharmacopoeia (BP) Reference Standard