missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Vitronectin, Human Purified Protein

Catalog No. CC080
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC080 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC080 Supplier MilliporeSigma™ Supplier No. CC080

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used to promote cell attachment, spreading, proliferation, and differentiation of normal and neoplastic cells

Specifications

Concentration 1 mg/mL
For Use With (Application) Positive control, Cell Culture
Formulation Liquid in 50 mM sodium phosphate, 100 mM NaCl, pH 7.4, containing no preservatives
Quantity 100 μg
Source Human plasma; tested negative for antibodies to HBsAg and HIV.
Product Type Vitronectin, Human Purified Protein
Content And Storage Stable 12 months at -70°C. Avoid repeated freeze/thaw cycles. 
Form Liquid
Purity or Quality Grade ≥90% by SDS-PAGE. Two bands in SDS-PAGE at 65 kd and 75 kd.