missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Millicoat™ Human Vitronectin Coated Strips (96-Wells)

Catalog No. ECM102
Change view
Click to view available options
Quantity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ECM102 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ECM102 Supplier MilliporeSigma™ Supplier No. ECM102

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Activity assay applications

  • Provided as 12 x 8-well removable strips in a plate frame for convenience and flexibility in designing assays
  • Wells in rows A to G have been coated with Human Vitronectin
  • Row H of each strip is coated with BSA which serves as negative assay control

Specifications

For Use With (Application) Bioactivity
Quantity 1 x 96-well Plate
Product Type Human Vitronectin Coated Strips
Content And Storage May be stored at 2° to 8°C in the foil pouch for at least 3 months