Learn More
MilliporeSigma™ Chemicon™ Millicoat™ Human Fibronectin Coated Strips (96-Wells)
Description
- Provided as 12 x 8-well removable strips in a plate frame for convenience and flexibility in designing assays
- Wells in rows A to G have been coated with Human Fibronectin
- Row H of each strip is coated with BSA which serves as negative assay control
Spécifications
Spécifications
| À utiliser avec (application) | Bioactivity |
| Quantité | 1 x 96-well Plate |
| Type de produit | Human Fibronectin Coated Strips |
| Contenu et stockage | May be stored at 2° to 8°C in the foil pouch for at least 3 months |