missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Chemicon™ Laminin, Mouse Purified
Spécifications
Spécifications
| Concentration | 1 mg/mL |
| For Use With (Application) | Cell Culture |
| Formulation | Liquid; in 0.05 M Tris-HCl, pH 7.4, containing 0.15 M NaCl. |
| Quantity | 1 mg |
| Source | Isolated from Engelbreth-Holm-Swarm (EHS) mouse sarcoma |
| Product Type | Laminin, Mouse Purified |
| Content And Storage | Store at -20°C for up to six months. |
| Form | Liquid |
| Purity or Quality Grade | >95% by SDS-PAGE |