missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Human Tenascin-C Purified Protein

Catalog No. CC065
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC065 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC065 Supplier MilliporeSigma™ Supplier No. CC065

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Plays active role in development of central nervous system and mesenchymal-derived organs

  • Exist in vasculature of tumors in adults
  • Function in cell adhesion
  • By SDS-PAGE analysis, the protein migrates around 280 to 300 kd

Specifications

Concentration 0.1 mg/mL
For Use With (Application) Positive control, Cell Culture
Quantity 100 μg
Product Type Human Tenascin-C Purified Protein
Content And Storage Maintain at -70°C for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.
Form Liquid
Purity or Quality Grade >97% pure by SDS-PAGE