missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Chemicon™ Human Plasma Fibronectin Purified Protein
Description
- Used as attachment factor
Spécifications
Spécifications
| Concentration | 1 mg/mL |
| À utiliser avec (application) | Cell Culture |
| Formule | Liquid in 150 mM NaCl, 10 mM sodium phosphate, pH 7.5, sterile filtered, containing no preservatives. |
| Plage de pH | 7.5 |
| Quantité | 5 mg |
| Sources | Human Donor Plasma |
| Type de produit | Human Plasma Fibronectin Purified Protein |
| Contenu et stockage | Maintain at 2° to 8°C for up to 6 months from date of receipt. Do not freeze. |
| Forme | Liquid |