missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Chemicon™ Human Laminin (Pepsinized) Purified Protein
Description
- Contains mixture of human laminins containing beta1 chain, principally laminin 1, 2, 3, 6, 8 and 10
- Preparation is immunologically and biologically identical to intact human laminin
- Runs as twin bands migrating at approximately 160 kd and 130 kd by SDS-PAGE under reducing conditions
Spécifications
Spécifications
| Concentration | 0.153 mg/mL |
| À utiliser avec (application) | Cell Culture |
| Quantité | 100 μg |
| Sources | Human placenta, tested negative for hepatitis B virus, hepatitis C virus (HCV), HIV-1, HIV-2, HTLV-1, HTLV-2, and Treponema pallidum. Handle as if potentially infectious. |
| Type de produit | Human Laminin (Pepsinized) Purified Protein |
| Contenu et stockage | Store at −20°C or -70°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. |
| Forme | Liquid |
| Grade de qualité ou de pureté | ≥95% by SDS-PAGE |