missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Chicken Extracellular Chondroitin Sulfate Proteoglycans

Numéro de catalogue. CC117
Modifier l'affichage
Click to view available options
Quantité:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC117 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC117 Fournisseur MilliporeSigma™ Code fournisseur CC117

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

For Cell Culture applications

  • Contains mixture of large, extracellular chondroitin sulfateproteoglycans isolated from embryonic chicken brain
  • Major components of this mixture are neurocan, phosphacan, versican, and aggrecan

Spécifications

Concentration 1 mg/mL
À utiliser avec (application) Cell Culture
Quantité 100 μg
Sources Isolated from whole 12-14 day old embryonic chicken brains.
Type de produit Chicken Extracellular Chondroitin Sulfate Proteoglycans
Contenu et stockage Store at -20°C for up to 12 months. Avoid repeated freeze/thaw.
Forme Liquid