Learn More
CHEMetrics CHEMets Dissolved Carbon Dioxide, Visual Water Testing Kit
Catalog No. 23291216
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
23291216 30
1 options
Catalog No. 23291216 Supplier CHEMetrics Supplier No. K1910
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Complete test kits contain 30 ampules of reagent, comparator(s), snap cup (holds water sample), instructions, and MSDS sheets. All components are housed in a sturdy, plastic case
Provides results in two minutes or less
Provide results in two minutes or less
  • Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
  • To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
  • In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
  • Comparators have two-year shelf life.
Specifications
Analytes Dissolved Carbon Dioxide
Includes Cardboard box, 30 ampoules of reagent with valve assemblies, titrettor, 25mL sample cup, instructions, activator solution
No. of Tests 30
Range 10 to 100 ppm
Test Method Caustic titrant with pH indicator
Type Chemets™ Water Testing Kit