Learn More
CHEMetrics CHEMets Iron Test Kit Refill
Catalog No. 13299213A
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13299213A 30
1 options
Catalog No. 13299213A Supplier CHEMetrics Supplier No. R6001
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Provides results in two minutes or less
This product contains 30 self-filling CHEMets ampules to refill kit K-6010 for the analysis of iron (total & soluble). R-6001 is compatible with C-6001 and C-6010 comparators.
Specifications
Analytes Iron
For Use With (Equipment) C-6001 and C-6010 comparators
Includes 30 self-filling CHEMets ampoules to refill kit K-6010
No. of Tests 30
Range 0 to 1 ppm and 1 to 10 ppm
Test Method Phenanthroline
Test Time 1 min.; Total 5 min.
Type CHEMets Refill Kit